siRNA Details
Gene Symbol: Gpx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022525.3 | NP_071970.2 | 5 | 698 | ||
GenBank mRNA | BC062227.1 | - | 5 | 674 | 6921653 | |
CV079715.1 | - | 4 | 437 | |||
FQ221037.1 | - | 5 | 670 | |||
FQ221279.1 | - | 5 | 651 | |||
FQ223139.1 | - | 4 | 413 | |||
FQ228485.1 | - | 5 | 670 | |||
FQ228545.1 | - | 5 | 724 | |||
FQ228679.1 | - | 5 | 670 | |||
FQ228689.1 | - | 5 | 670 | |||
FQ228690.1 | - | 5 | 683 | |||
FQ228694.1 | - | 5 | 650 | |||
FQ228697.1 | - | 5 | 670 | |||
FQ228699.1 | - | 5 | 670 | |||
FQ228705.1 | - | 5 | 668 | |||
FQ228712.1 | - | 5 | 670 | |||
FQ228719.1 | - | 5 | 670 | |||
FQ228737.1 | - | 5 | 670 | |||
FQ228788.1 | - | 5 | 670 | |||
FQ228802.1 | - | 5 | 670 | |||
FQ228803.1 | - | 5 | 667 | |||
FQ228804.1 | - | 5 | 670 | |||
FQ228807.1 | - | 5 | 671 | |||
FQ228808.1 | - | 5 | 670 | |||
FQ228824.1 | - | 5 | 670 | |||
FQ228834.1 | - | 5 | 672 | |||
FQ228835.1 | - | 5 | 670 | |||
FQ228845.1 | - | 5 | 670 | |||
FQ228866.1 | - | 5 | 670 | |||
FQ228878.1 | - | 5 | 670 | |||
FQ228879.1 | - | 5 | 670 | |||
FQ228886.1 | - | 5 | 670 | |||
FQ228899.1 | - | 5 | 670 | |||
FQ228911.1 | - | 5 | 670 | |||
FQ228914.1 | - | 5 | 670 | |||
FQ228921.1 | - | 5 | 670 | |||
FQ228934.1 | - | 5 | 672 | |||
FQ228945.1 | - | 5 | 670 | |||
FQ228947.1 | - | 5 | 670 | |||
FQ228960.1 | - | 5 | 670 | |||
FQ228965.1 | - | 5 | 670 | |||
FQ228968.1 | - | 5 | 670 | |||
FQ228969.1 | - | 5 | 669 | |||
FQ228970.1 | - | 5 | 670 | |||
FQ228976.1 | - | 5 | 670 | |||
FQ229118.1 | - | 5 | 670 | |||
FQ229125.1 | - | 5 | 670 | |||
FQ230052.1 | - | 4 | 413 |
