siRNA Details
Gene Symbol: Bad | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022698.1 | NP_073189.1 | 4 | 573 | ||
XM_006230898.2 | XP_006230960.1 | 4 | 471 | |||
GenBank mRNA | AF003523.1 | - | 4 | 573 | ||
AF031227.1 | - | 4 | 611 | |||
AF279910.1 | - | 4 | 573 |
