siRNA Details
Gene Symbol: Asic1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024154.2 | NP_077068.1 | 7 | 1146 | ||
XM_006257378.2 | XP_006257440.1 | 5 | 1471 | |||
XM_006257381.2 | XP_006257443.1 | 7 | 1431 | |||
XM_008765793.1 | XP_008764015.1 | 5 | 1475 | |||
XM_008765794.1 | XP_008764016.1 | 7 | 1434 | |||
XM_008765795.1 | XP_008764017.1 | 7 | 1087 | |||
XM_008765796.1 | XP_008764018.1 | 7 | 1431 | |||
GenBank mRNA | AB049451.1 | - | 5 | 737 | ||
AJ006519.1 | - | 5 | 1192 | |||
AJ309926.1 | - | 5 | 1196 |
