siRNA Details
Gene Symbol: Rabep2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030585.1 | NP_085074.1 | 2, 3 | 293 | ||
GenBank mRNA | BC089795.1 | - | 2, 3 | 279 | 7375879 | |
U34932.1 | - | 2, 3 | 293 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_030566, AK089938, AK157832, BU514356, BC015287, and others.
Gene Ontology Categories:
