siRNA Details
Gene Symbol: Scn1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030875.1 | NP_110502.1 | 23, 24 | 4592 | ||
XM_008761923.1 | XP_008760145.1 | 23, 24 | 4612 | |||
XM_008761924.1 | XP_008760146.1 | 23, 24 | 4520 | |||
XM_008761925.1 | XP_008760147.1 | 23, 24 | 4428 | |||
XM_008761926.1 | XP_008760148.1 | 23, 24 | 4611 | |||
XM_008761927.1 | XP_008760149.1 | 23, 24 | 4578 | |||
XM_008761928.1 | XP_008760150.1 | 23, 24 | 4576 | |||
XM_008761929.1 | XP_008760151.1 | 22, 23 | 4329 | |||
GenBank mRNA | FQ212107.1 | - | 8, 9 | 1119 | ||
M22253.1 | - | 23, 24 | 4592 | |||
X03638.1 | - | 23, 24 | 4592 |
