siRNA Details
Gene Symbol: Vim | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031140.1 | NP_112402.1 | 5 | 1068 | ||
GenBank mRNA | BC061847.1 | - | 5 | 1108 | 5621065 | |
FQ220610.1 | - | 3 | 371 | |||
FQ229240.1 | - | 3 | 375 | |||
X62952.1 | - | 5 | 1068 |
