siRNA Details
Gene Symbol: Vim | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031140.1 | NP_112402.1 | 1 | 361 | ||
GenBank mRNA | BC061847.1 | - | 1 | 401 | 5621065 | |
M84481.1 | - | 1 | 27 | |||
X62952.1 | - | 1 | 361 |

More Information
Other Species Targeted

Gene Ontology Categories:
