siRNA Details
Gene Symbol: Grb7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053403.2 | NP_445855.1 | 8 | 1280 | ||
XM_006247432.2 | XP_006247494.1 | 8 | 3249 | |||
XM_006247433.2 | XP_006247495.1 | 8 | 1132 | |||
XM_006247434.2 | XP_006247496.1 | 8 | 1532 | |||
XM_008768125.1 | XP_008766347.1 | 8 | 1066 | |||
GenBank mRNA | AF190121.1 | - | 8 | 881 | ||
BC081757.1 | - | 8 | 1280 | 7109771 |
