siRNA Details
Gene Symbol: Ran | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053439.1 | NP_445891.1 | 4 | 237 | ||
XM_006249285.2 | XP_006249347.1 | 4 | 224 | |||
GenBank mRNA | AF306457.1 | - | 4 | 237 | ||
BC059123.1 | - | 4 | 252 | 6920235 | ||
FQ220083.1 | - | 4 | 220 | |||
FQ220326.1 | - | 4 | 220 | |||
FQ220799.1 | - | 4 | 265 | |||
FQ229342.1 | - | 4 | 189 | |||
FQ229588.1 | - | 4 | 243 | |||
FQ229761.1 | - | 4 | 228 | |||
FQ231051.1 | - | 3 | 232 | |||
FQ233248.1 | - | 4 | 191 |
