siRNA Details
Gene Symbol: Tgfb1i1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191840.1 | NP_001178769.1 | 9 | 950 | ||
XM_006230349.2 | XP_006230411.1 | 9 | 1800 | |||
XM_006230350.2 | XP_006230412.1 | 9 | 922 | |||
XM_008759920.1 | XP_008758142.1 | 8 | 2007 | |||
XM_008759921.1 | XP_008758143.1 | 9 | 1007 | |||
GenBank mRNA | AF314960.1 | - | 5 | 530 |
