siRNA Details
Gene Symbol: Bmp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191856.2 | NP_001178785.1 | 3 | 1092 | ||
GenBank mRNA | AF100787.1 | - | 3 | 337 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Bmp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191856.2 | NP_001178785.1 | 3 | 1092 | ||
GenBank mRNA | AF100787.1 | - | 3 | 337 |