siRNA Details
Gene Symbol: Gata3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133293.1 | NP_579827.1 | 4, 5 | 1038 | ||
XM_006254253.2 | XP_006254315.1 | 5, 6 | 1804 | |||
XM_008771893.1 | XP_008770115.1 | 5, 6 | 1748 | |||
XM_008771894.1 | XP_008770116.1 | 5, 6 | 3338 | |||
GenBank mRNA | AY024364.1 | - | 4, 5 | 1038 |
