siRNA Details
Gene Symbol: Dut | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040271.1 | NP_001035361.1 | 6 | 496 | ||
NM_053592.2 | NP_446044.1 | 7 | 616 | |||
XM_008762188.1 | XP_008760410.1 | 6 | 583 | |||
GenBank mRNA | AI705003.1 | - | 1 | 327 | ||
CB325379.1 | - | 6 | 510 | |||
FQ220451.1 | - | 651 | ||||
FQ233848.1 | - | 2 | 116 | |||
U64030.2 | - | 7 | 616 |
