siRNA Details
Gene Symbol: Slc9a3r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053811.3 | NP_446263.2 | 7 | 1020 | ||
XM_006245893.2 | XP_006245955.1 | 6 | 1393 | |||
GenBank mRNA | AF259898.1 | - | 7 | 879 | ||
AF294257.1 | - | 7 | 1107 |
