siRNA Details
Gene Symbol: Cyth2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053911.2 | NP_446363.1 | 8 | 900 | ||
XM_006228964.2 | XP_006229026.1 | 8 | 918 | |||
XM_008759304.1 | XP_008757526.1 | 7 | 1391 | |||
GenBank mRNA | FQ229744.1 | - | 7 | 621 | ||
U83896.1 | - | 8 | 904 |
