siRNA Details
Gene Symbol: Dynll2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080697.2 | NP_542428.1 | 2, 3 | 387 | ||
XM_006247077.2 | XP_006247139.1 | 2, 3 | 166 | |||
XM_008767940.1 | XP_008766162.1 | 2, 3 | 157 | |||
GenBank mRNA | AY034383.1 | - | 2, 3 | 281 | ||
BC061874.1 | - | 2, 3 | 387 | 5598327 |
