siRNA Details
Gene Symbol: Prph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012633.2 | NP_036765.2 | 7, 8 | 1294 | ||
XM_006257321.2 | XP_006257383.1 | 6, 7 | 2173 | |||
XM_006257322.2 | XP_006257384.1 | 7, 8 | 2075 | |||
XM_008765669.1 | XP_008763891.1 | 6, 7 | 2175 | |||
GenBank mRNA | AF031878.1 | - | 7, 8 | 1287 | ||
BC100656.1 | - | 7, 8 | 1294 | 7936222 | ||
BP496275.1 | - | 2, 3 | 463 |
