siRNA Details
Gene Symbol: Aqp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012779.1 | NP_036911.1 | 1 | 241 | ||
GenBank mRNA | BC078904.1 | - | 1 | 235 | ||
BC092572.1 | - | 1 | 230 | 7122974 | ||
BC110045.1 | - | 1 | 246 | 7100092 | ||
U16245.1 | - | 1 | 241 |
