siRNA Details
Gene Symbol: Scn2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012877.1 | NP_037009.1 | 3 | 540 | ||
XM_006242895.2 | XP_006242957.1 | 3 | 492 | |||
GenBank mRNA | U37026.1 | - | 3 | 540 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Scn2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012877.1 | NP_037009.1 | 3 | 540 | ||
XM_006242895.2 | XP_006242957.1 | 3 | 492 | |||
GenBank mRNA | U37026.1 | - | 3 | 540 |