siRNA Details
Gene Symbol: G6pc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013098.2 | NP_037230.2 | 1 | 174 | ||
GenBank mRNA | BC090067.1 | - | 1 | 174 | 7109006 | |
D78592.1 | - | 1 | 186 | |||
L37333.1 | - | 1 | 138 | |||
U07993.1 | - | 1 | 136 |
