siRNA Details
Gene Symbol: Il7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013110.2 | NP_037242.2 | 2 | 134 | ||
XM_006232139.2 | XP_006232201.1 | 2 | 2980 | |||
XM_008760860.1 | XP_008759082.1 | 2 | 2980 | |||
GenBank mRNA | AF010464.1 | - | 2 | 143 | ||
AF367210.1 | - | 2 | 134 | |||
FQ226544.1 | - | 704 | ||||
FQ226549.1 | - | 704 |
