siRNA Details
Gene Symbol: Sorbs3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005762.1 | NP_001005762.1 | 17 | 1557 | ||
XM_006252241.2 | XP_006252303.1 | 18 | 3241 | |||
XM_006252242.1 | XP_006252304.1 | 17 | 1648 | |||
XM_006252244.2 | XP_006252306.1 | 16 | 1876 | |||
XM_006252246.1 | XP_006252308.1 | 17 | 1593 | |||
XM_006252248.1 | XP_006252310.1 | 5 | 441 | |||
XM_008770819.1 | XP_008769041.1 | 15 | 3350 | |||
GenBank mRNA | AF439378.1 | - | 16 | 1373 | ||
BC083666.1 | - | 17 | 1557 | 7135547 |
