siRNA Details
Gene Symbol: Emc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105806.1 | NP_001099276.1 | 1 | 195 | ||
XM_006246669.2 | XP_006246731.1 | 2 | 298 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Emc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105806.1 | NP_001099276.1 | 1 | 195 | ||
XM_006246669.2 | XP_006246731.1 | 2 | 298 |