siRNA Details
Gene Symbol: Mnt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105807.1 | NP_001099277.1 | 6 | 2005 | ||
XM_008767959.1 | XP_008766181.1 | 6 | 12650 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mnt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105807.1 | NP_001099277.1 | 6 | 2005 | ||
XM_008767959.1 | XP_008766181.1 | 6 | 12650 |