siRNA Details
Gene Symbol: Zdhhc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039099.1 | NP_001034188.1 | 4 | 457 | ||
XM_008772485.1 | XP_008770707.1 | 4 | 2233 | |||
XM_008772486.1 | XP_008770708.1 | 4 | 2232 | |||
GenBank mRNA | AY886520.1 | - | 4 | 457 | ||
BC160816.1 | - | 3 | 376 |
