siRNA Details
Gene Symbol: Psmb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025637.1 | NP_001020808.1 | 7 | 796 | ||
GenBank mRNA | BC098835.1 | - | 7 | 796 | 7378841 | |
FQ217478.1 | - | 3 | 339 | |||
FQ226463.1 | - | 7 | 789 | |||
FQ228226.1 | - | 7 | 776 | |||
FQ229295.1 | - | 7 | 794 | |||
FQ233484.1 | - | 7 | 806 | |||
FQ234080.1 | - | 7 | 798 | |||
FQ235075.1 | - | 7 | 779 |
