siRNA Details
Gene Symbol: Psmb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025637.1 | NP_001020808.1 | 5 | 551 | ||
GenBank mRNA | BC098835.1 | - | 5 | 551 | 7378841 | |
FQ217478.1 | - | 1 | 94 | |||
FQ226463.1 | - | 5 | 544 | |||
FQ228226.1 | - | 5 | 531 | |||
FQ229295.1 | - | 5 | 549 | |||
FQ229779.1 | - | 604 | ||||
FQ233484.1 | - | 5 | 561 | |||
FQ234080.1 | - | 5 | 553 | |||
FQ235075.1 | - | 5 | 534 |
