siRNA Details
Gene Symbol: Psmb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025637.1 | NP_001020808.1 | 3 | 302 | ||
GenBank mRNA | BC098835.1 | - | 3 | 302 | 7378841 | |
FQ226463.1 | - | 3 | 295 | |||
FQ228226.1 | - | 3 | 283 | |||
FQ229295.1 | - | 3 | 300 | |||
FQ229779.1 | - | 355 | ||||
FQ233484.1 | - | 3 | 312 | |||
FQ234080.1 | - | 3 | 304 | |||
FQ235075.1 | - | 3 | 285 |
