siRNA Details
Gene Symbol: Smoc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106215.2 | NP_001099685.1 | 11 | 1418 | ||
XM_006227958.2 | XP_006228020.1 | 11 | 1508 | |||
GenBank mRNA | BC168953.1 | - | 11 | 1418 | 7109706 |
