siRNA Details
Gene Symbol: Mesp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106273.1 | NP_001099743.1 | 1 | 736 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mesp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106273.1 | NP_001099743.1 | 1 | 736 |