siRNA Details
Gene Symbol: Pla2g5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017174.1 | NP_058870.1 | 5 | 575 | ||
XM_006239139.2 | XP_006239201.1 | 6 | 1374 | |||
XM_006239140.2 | XP_006239202.1 | 6 | 1953 | |||
XM_006239141.2 | XP_006239203.1 | 5 | 497 | |||
XM_008764209.1 | XP_008762431.1 | 7 | 1488 | |||
XM_008764210.1 | XP_008762432.1 | 5 | 2595 | |||
XM_008764211.1 | XP_008762433.1 | 4 | 610 | |||
GenBank mRNA | AY651028.1 | - | 5 | 442 | ||
BC085745.1 | - | 5 | 600 | 7192819 | ||
U03763.1 | - | 5 | 575 |
