siRNA Details
Gene Symbol: Lgals12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106333.1 | NP_001099803.1 | 2 | 574 | ||
XM_006230763.2 | XP_006230825.1 | 3 | 1141 | |||
XM_006230764.2 | XP_006230826.1 | 3 | 404 | |||
XM_008760110.1 | XP_008758332.1 | 3 | 1353 | |||
XM_008760111.1 | XP_008758333.1 | 3 | 1337 | |||
XM_008760112.1 | XP_008758334.1 | 2 | 503 |
