siRNA Details
Gene Symbol: Adra1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024483.1 | NP_077809.1 | 1 | 857 | ||
GenBank mRNA | L31771.1 | - | 1 | 857 | ||
M60654.1 | - | 1 | 857 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
