siRNA Details
Gene Symbol: Nus1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164157.1 | NP_001157629.1 | 3, 4 | 781 | ||
GenBank mRNA | BQ782237.1 | - | 2, 3 | 302 | ||
CK363676.1 | - | 3, 4 | 300 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nus1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164157.1 | NP_001157629.1 | 3, 4 | 781 | ||
GenBank mRNA | BQ782237.1 | - | 2, 3 | 302 | ||
CK363676.1 | - | 3, 4 | 300 |