siRNA Details
Gene Symbol: Nus1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164157.1 | NP_001157629.1 | 1 | 487 | ||
GenBank mRNA | CB720603.1 | - | 1 | 487 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_030250, AK167939, AK155620, BC003223, BC018372, and others.
Gene Ontology Categories:
