siRNA Details
Gene Symbol: Nus1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164157.1 | NP_001157629.1 | 2 | 594 | ||
GenBank mRNA | BQ782237.1 | - | 4 | 489 | ||
CK363676.1 | - | 2 | 113 |

More Information
Other Species Targeted

Gene Ontology Categories:
