siRNA Details
Gene Symbol: Atn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CB702622.1 | - | 2 | 146 | ||
U31777.1 | - | 5 | 634 |
Gene Symbol: LOC100911672 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750658.3 | XP_003750706.1 | 5 | 621 | ||
XM_006244637.2 | XP_006244699.1 | 5 | 607 | |||
XM_008766913.1 | XP_008765135.1 | 5 | 589 |
