siRNA Details
Gene Symbol: Rbm8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271138.1 | NP_001258067.1 | 4, 5 | 401 | ||
XM_006232956.2 | XP_006233018.1 | 4, 5 | 399 | |||
GenBank mRNA | DQ359102.1 | - | 4, 5 | 353 | ||
FQ210627.1 | - | 4, 5 | 374 |
