siRNA Details
Gene Symbol: LOC100911617 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008761447.1 | XP_008759669.1 | 3331 |
Gene Symbol: Prmt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106466.1 | NP_001099936.1 | 1 | 1236 |
