siRNA Details
Gene Symbol: Eci1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017306.4 | NP_059002.2 | 6 | 688 | ||
GenBank mRNA | BC078705.1 | - | 6 | 688 | 7129389 | |
FQ213384.1 | - | 6 | 696 | |||
FQ218351.1 | - | 6 | 689 | |||
M61112.1 | - | 6 | 677 | |||
X61184.1 | - | 6 | 699 |
