siRNA Details
Gene Symbol: RGD1566368 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729986.4 | XP_002730032.2 | 4, 5 | 1599 | ||
XM_006244213.2 | XP_006244275.1 | 4, 5 | 1219 | |||
XM_008766730.1 | XP_008764952.1 | 5, 6 | 1110 | |||
XM_008766731.1 | XP_008764953.1 | 4, 5 | 1333 |
