siRNA Details
Gene Symbol: Pdzd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106945.1 | NP_001100415.1 | 5 | 483 | ||
XM_006257068.2 | XP_006257130.1 | 5 | 389 | |||
XM_006257070.2 | XP_006257132.1 | 4 | 324 | |||
GenBank mRNA | BC168740.1 | - | 4 | 247 | 6918646 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_028303, AK012624, AK032327, BC004608, AK007454.
Gene Ontology Categories:
