siRNA Details
Gene Symbol: Eif1ax | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106963.1 | NP_001100433.1 | 4 | 441 | ||
GenBank mRNA | BC168938.1 | - | 4 | 314 | 7097069 | |
FQ217925.1 | - | 4 | 406 | |||
FQ221854.1 | - | 4 | 420 | |||
FQ222873.1 | - | 4 | 439 |
