siRNA Details
Gene Symbol: Eif1ax | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106963.1 | NP_001100433.1 | 3 | 374 | ||
GenBank mRNA | BC168938.1 | - | 3 | 247 | 7097069 | |
FQ217925.1 | - | 3 | 339 | |||
FQ221854.1 | - | 3 | 353 | |||
FQ222873.1 | - | 3 | 372 |
