siRNA Details
Gene Symbol: Mrpl38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009369.2 | NP_001009369.2 | 4 | 573 | ||
XM_008768372.1 | XP_008766594.1 | 4 | 1361 | |||
GenBank mRNA | BC087096.1 | - | 4 | 570 | 7191805 | |
CO567099.1 | - | 4 | 573 |
