siRNA Details
Gene Symbol: Ccsap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107441.1 | NP_001100911.1 | 2 | 488 | ||
XM_006255811.2 | XP_006255873.1 | 3 | 988 | |||
GenBank mRNA | FQ211944.1 | - | 2 | 262 | ||
FQ212467.1 | - | 2 | 261 |
