siRNA Details
Gene Symbol: Klk12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107508.1 | NP_001100978.1 | 3 | 490 | ||
XM_008759351.1 | XP_008757573.1 | 5 | 570 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Klk12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107508.1 | NP_001100978.1 | 3 | 490 | ||
XM_008759351.1 | XP_008757573.1 | 5 | 570 |