siRNA Details
Gene Symbol: Rhog | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037195.1 | NP_001032272.1 | 2 | 588 | ||
XM_006229882.2 | XP_006229944.1 | 3 | 729 | |||
XM_006229883.2 | XP_006229945.1 | 2 | 835 | |||
GenBank mRNA | BC107943.1 | - | 2 | 588 | 7389075 | |
FQ212590.1 | - | 2 | 628 | |||
FQ232547.1 | - | 2 | 631 |
