siRNA Details
Gene Symbol: Rab25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107687.2 | NP_001101157.2 | 4 | 726 | ||
GenBank mRNA | BC168745.1 | - | 4 | 723 | 6920264 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rab25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107687.2 | NP_001101157.2 | 4 | 726 | ||
GenBank mRNA | BC168745.1 | - | 4 | 723 | 6920264 |