siRNA Details
Gene Symbol: Fam110a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014050.2 | NP_001014072.2 | 2 | 1214 | ||
NM_001244768.1 | NP_001231697.1 | 2 | 1095 | |||
NM_001244769.1 | NP_001231698.1 | 2 | 1167 | |||
NM_001244770.1 | NP_001231699.1 | 2 | 1014 | |||
XM_006235231.2 | XP_006235293.1 | 2 | 1467 | |||
XM_006235234.2 | XP_006235296.1 | 3 | 1202 | |||
GenBank mRNA | BC079251.1 | - | 2 | 1214 | 7133086 | |
CB556608.1 | - | 1 | 671 | |||
CB766675.1 | - | 1 | 327 | |||
JF912497.1 | - | 1 | 825 |
